Learn More
SiliCycle™ SiliaMetS™ E-PAK™ C-VW Activated Carbon
Our C-VW is a virgin wood-based catalytic activated carbon.
Supplier:  SiliCycle™ CVW20KG
 View more versions of this product
Catalog No. 01-182-167
Add to cart
Description
Description
SiliaCarb VW is a carbon with superior catalytic activity and ultra high purity for water purification. Highly effective for removing chloramine and chlorine.
  • Virgin wood-based
  • Catalytic activated
Specifications
Specifications
Black
20 kg
E-PAK
5 to 9
Powder
Removal of precious metal catalysts, pigments and reaction contaminants