Learn More
SiliCycle™ SiliaMetS™ Metal Scavengers AMPA, 40 to 63 μm, 60 Å
Our AMPA is a powerful scavenger for the removal of Al, Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Mg, Nd, Ni, Pm, Pr, Sb, Sm, Tb, Tm, V and Yb.
$45.45 - $3520.00
Specifications
Chemical Name or Material | Bulk Functionalized Irregular Silica Gel |
---|---|
Physical Form | Irregular |
Packaging | HDPE Bottle |
Product Line | SiliaMetS™ |
Density | 0.707 g/mL |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-184-052
|
SiliCycle™
R85130B5G |
5 g |
Each for $45.45
|
|
01-184-053
|
SiliCycle™
R85130B10G |
10 g |
Each for $74.10
|
|
01-184-054
|
SiliCycle™
R85130B25G |
25 g |
Each for $147.00
|
|
01-184-055
|
SiliCycle™
R85130B50G |
50 g |
Each for $276.50
|
|
01-184-056
|
SiliCycle™
R85130B100G |
100 g |
Each for $440.00
|
|
01-184-057
|
SiliCycle™
R85130B250G |
250 g |
Each for $1,036.00
|
|
01-184-058
|
SiliCycle™
R85130B500G |
500 g |
Each for $1,955.00
|
|
01-184-059
|
SiliCycle™
R85130B1KG |
1 kg |
Each for $3,520.00
|
|
Description
SiliaMetS™ AMPA is an aminomethylalkylphosphonic acid ligand known for its excellent metal-bonding properties. It is particularly efficient to remove Al, Sb, Ni, La from reaction intermediates or final APIs. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. It offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Best for Al, Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Mg, Nd, Ni, Pm, Pr, Sb, Sm, Tb, Tm, V and Yb
- Also good for Co, Cu, Fe, Mg and Zn
- Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
Bulk Functionalized Irregular Silica Gel | |
HDPE Bottle | |
0.707 g/mL |
Irregular | |
SiliaMetS™ |
Safety and Handling
Recommended Storage : Keep Dry
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.