Learn More
SiliCycle™ SiliaMetS™ Metal Scavengers DEAM, 40 to 63 μm, 60 Å
DEAM is a powerful metal scavenger designed for trapping Ag, Fe, Sn, Ti and Zn, as well as acting as an organic scavenger for the removal of boronic acids.
Supplier: SiliCycle™ R54430B500G
Description
SiliaMetS™ DEAM is a versatile Metal Scavenger designed to remove trace metal of Ti, Zn, Fe and Ag (best for: Ag, Fe, Sn, Ti and Zn). You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Great for a variety of formats for all purifications scales, from laboratory to process to plant scales. The DEAM is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and offers great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with a sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
Bulk Functionalized Irregular Silica Gel | |
500 g | |
SiliaMetS™ |
Irregular | |
HDPE Bottle | |
0.691 g/mL |
Safety and Handling
Recommended Storage : Keep Cool (<8°C) and Dry
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.