Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaMetS™ Metal Scavengers DEAM, 40 to 63 μm, 60 Å

DEAM is a powerful metal scavenger designed for trapping Ag, Fe, Sn, Ti and Zn, as well as acting as an organic scavenger for the removal of boronic acids.

$107.10 - $5070.00

Specifications

Chemical Name or Material Bulk Functionalized Irregular Silica Gel
Physical Form Irregular
Packaging HDPE Bottle
Product Line SiliaMetS™
Density 0.691 g/mL
View More Specs

Products 8
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-183-989
View Documents
SiliCycle™
R54430B10G
10 g
Each for $107.10
Add to Cart
 
01-183-990
View Documents
SiliCycle™
R54430B25G
25 g
Each for $219.50
Add to Cart
 
01-183-991
View Documents
SiliCycle™
R54430B50G
50 g
Each for $411.50
Add to Cart
 
01-183-992
View Documents
SiliCycle™
R54430B100G
100 g
Each for $634.50
Add to Cart
 
01-183-993
View Documents
SiliCycle™
R54430B250G
250 g
Each for $1,591.00
Add to Cart
 
01-183-994
View Documents
SiliCycle™
R54430B500G
500 g
Each for $2,840.00
Add to Cart
 
01-183-995
View Documents
SiliCycle™
R54430B1KG
1 kg
Each for $5,070.00
Add to Cart
 
01-183-988
View Documents
SiliCycle™
R54430B5G
5 g Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
Description

Description

SiliaMetS™ DEAM is a versatile Metal Scavenger designed to remove trace metal of Ti, Zn, Fe and Ag (best for: Ag, Fe, Sn, Ti and Zn). You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Great for a variety of formats for all purifications scales, from laboratory to process to plant scales. The DEAM is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and offers great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with a sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

Bulk Functionalized Irregular Silica Gel
HDPE Bottle
0.691 g/mL
Irregular
SiliaMetS™
Safety and Handling

Safety and Handling

Recommended Storage : Keep Cool (<8°C) and Dry

SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit