Learn More
SiliCycle™ SiliaMetS™ Metal Scavengers Diamine (PSA), 40 to 63 μm, 60 Å
Our Diamine is a powerful metal scavenger for trapping Cr, Pd, Pt, W and Zn, and also Cd, Co, Cu, Fe, Hg, Ni, Pb, Ru and Sc.
$42.55 - $3120.00
Specifications
Chemical Name or Material | Bulk Functionalized Irregular Silica Gel |
---|---|
Physical Form | Irregular |
Packaging | HDPE Bottle |
Product Line | SiliaMetS™ |
Density | 0.728 g/mL |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-183-964
|
SiliCycle™
R49030B5G |
5 g |
Each for $42.55
|
|
01-183-965
|
SiliCycle™
R49030B10G |
10 g |
Each for $65.90
|
|
01-183-966
|
SiliCycle™
R49030B25G |
25 g |
Each for $130.20
|
|
01-183-967
|
SiliCycle™
R49030B50G |
50 g |
Each for $245.00
|
|
01-183-968
|
SiliCycle™
R49030B100G |
100 g |
Each for $403.50
|
|
01-183-969
|
SiliCycle™
R49030B250G |
250 g |
Each for $945.00
|
|
01-183-970
|
SiliCycle™
R49030B500G |
500 g |
Each for $1,732.00
|
|
01-183-971
|
SiliCycle™
R49030B1KG |
1 kg |
Each for $3,120.00
|
|
Description
Also known for their electrophile scavenging efficiencies and their base reagent qualities, SiliaMetS™ Amine, Diamine and Triamine have also proven to be very useful for the scavenging of the following metals: Pd, Pt, Cr, W and Zn. SiliaBond™ Diamine acts as a Metal Scavenger and an Organic Scavenger for Electrophiles and Acids. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Our Diamine offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Best for Cr, Pd, Pt, W and Zn
- Also good for Cd, Co, Cu, Fe, Hg, Ni, Pb, Ru and Sc
- Covalent Binding for Acyl Chlorides, Aldehydes, Anhydrides, Chloroformates, Isocyanates, Ketones and
Sulfonyl Chlorides - Ionic Binding and Catch and Release for Acids and Acidic Phenols
- Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
Bulk Functionalized Irregular Silica Gel | |
HDPE Bottle | |
0.728 g/mL |
Irregular | |
SiliaMetS™ |
Safety and Handling
Recommended Storage : Keep Cool (<8°C) and Dry
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.