Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaMetS™ Metal Scavengers TAAcONa nec, 40 to 63 μm, 60 Å

Our TAAcONa Metal Scavenger is a version of EDTA in its sodium salt form powerful for trapping Ca, Cd, Cs, Cu, Fe, Ir, La, Li, Mg, Ni, Os, Rh, Sc and Sn.

Supplier:  SiliCycle™ R69230B10G

 View more versions of this product

Catalog No. 01-184-013


Add to cart

Description

Description

SiliaMetS™ TAAcOH and TAAcONa are supported versions of EDTA in their acid and sodium salt forms. These two products are effective Metal Scavengers for Ca, Mg, Li, Ir, Cs, Os, Sn, Pd, Ni and Cu. SiliaMetS ™ TAAcONa is useful for metals in higher oxidation states (≥2) (Best for: Ca, Cd, Cs, Cu, Fe, Ir, La, Li, Mg, Ni, Os, Rh, Sc and Sn; also good for: Cr, Pd, Ru and Zn)

  • Only need to filter out the scavenger once it has trapped the residual metal or organic impurity
  • Great variety of formats for all purifications scales, from laboratory to process to plant scales
  • Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
  • Great compatibility with a myriad of experimental conditions, solvents, pH and temperatures
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

Bulk Functionalized Irregular Silica Gel
10 g
SiliaMetS™
Irregular
HDPE Bottle
0.712 g/mL
Safety and Handling

Safety and Handling

Recommended Storage : Keep Dry

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit