Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaMetS™ Metal Scavengers Triamine (TRI), 40 to 63 μm, 60 Å

Our Triamine is a powerful metal scavenger for trapping Cr, Pd, Pt, W and Zn, and also Ag, Cd, Co, Cu, Fe, Hg, Ni, Os, Pb, Rh, Ru and Sc.

Supplier:  SiliCycle™ R48030B1KG

 View more versions of this product

Catalog No. 01-183-963


Add to Cart

Description

Description

Also known for their electrophile scavenging efficiencies and their base reagent qualities, SiliaMetS™ Amine, Diamine and Triamine have also proven to be very useful for the scavenging of the following metals: Pd, Pt, Cr, W and Zn. SiliaBond™ Triamine acts as a Metal Scavenger and an Organic Scavenger for Electrophiles and Acids. You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. Our Triamine offers a great variety of formats for all purifications scales, from laboratory to process to plant scales, as well as great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.

  • Best for Cr, Pd, Pt, W and Zn
  • Also good for Ag, Cd, Co, Cu, Fe, Hg, Ni, Os, Pb, Rh, Ru and Sc
  • Covalent Binding for Acyl Chlorides, Aldehydes, Anhydrides, Chloroformates, Isocyanates, Ketones and Sulfonyl Chlorides
  • Ionic Binding and Catch and Release for Acids and Acidic Phenols
  • Successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment
  • Strong chemical, physical, thermal and mechanical stability
  • Made in accordance with sustainable process chemistry and green metrics
  • No hazards; no safety precautions needed
Specifications

Specifications

Bulk Functionalized Irregular Silica Gel
1 kg
SiliaMetS™
Irregular
HDPE Bottle
0.736 g/mL
Safety and Handling

Safety and Handling

Recommended Storage : Keep Cool (<8°C) and Dry

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit