Learn More
SiliCycle™ SiliaPlate™ HPTLC Plates, Glass-backed, Cyano Bound Silica
Glass TLC plates exhibiting high chemical resistance, high heating stability and charring resistance for quick and inexpensive separations.
Supplier:  SiliCycle™ TLGR38011B303
 View more versions of this product
Catalog No. 01-184-474
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
SilicaPlate™ Glass-backed HPTLC Plates are made with irregular particle shape and exhibit high chemical resistance.
  • Rigid plates
  • High heating stability and charring resistance
  • Transparent
  • Endcapped
  • Cyano-bound silica
Specifications
Specifications
Glass
10 to 12 μm
Glass-backed TLC Plates
TLC Analysis
Cyano (CN, Nitrile)
20 x 20 cm
Glass
25 Pk.
F254
SiliaPlate
150 um