Learn More
SiliCycle™ SiliaPlate™ HPTLC Plates, Glass-backed, Silica C2
Glass TLC plates exhibiting high chemical resistance, high heating stability and charring resistance for quick and inexpensive separations.
Supplier:  SiliCycle™ TLGR32611B203
 View more versions of this product
Catalog No. 01-184-466
Add to cart
Description
Description
SilicaPlate™ Glass-backed HPTLC Plates are made with irregular particle shape and exhibit high chemical resistance.
  • Rigid plates
  • High heating stability and charring resistance
  • Transparent
  • Endcapped
  • Silica C2
Specifications
Specifications
Glass
10 to 12 μm
Glass-backed TLC Plates
TLC Analysis
C2 (Dimethyl)
10 x 10 cm
Glass
25 Pk.
F254
SiliaPlate
150 μm