Learn More
SiliCycle™ SiliaPlate™ Preparative TLC Plates, Glass-Backed, Neutral Alumina
Glass preparative TLC plates designed for quantitative and qualitative purification of small quantities of a sample directly onto the TLC plate.
Supplier:  SiliCycle™ TLGAUT0337B341N
 View more versions of this product
Catalog No. 01-184-103
Add to cart
Description
Description
SiliaPlate™ Preparative TLC Plates are glass-backed and for irregular particle shape.
  • Rigid plates
  • High chemical resistance
  • High heating stability and charring resistance
  • Transparent
  • Neutral alumina array
Specifications
Specifications
Glass
15 Pk.
No
SiliaPlate
1000 μm
Glass
Glass-backed TLC Plates
Preparative TLC
Neutral Alumina (Aluminum Oxide, Al2O3)
20 x 20 cm