Learn More
SiliCycle™ SiliaPlate™ TLC CZ Plates; Glass-backed, Silica with Preadsorbent Zone
Glass TLC plates with concentrating zone to ensure sample integrity and improved resolution of spots, as well as to prevent crossover.
Supplier:  SiliCycle™ TLGZR10011B723
 View more versions of this product
Catalog No. 01-184-489
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
SiliaPlate™ Glass-backed TLC Plates with preadsorbent zone provide irregular particle shape.
  • Rigid
  • High chemical resistance
  • High heating stability and charring resistance
  • Transparent
Specifications
Specifications
Glass
10 to 12 μm
Glass-backed TLC Plates
TLC Analysis
Yes
250 μm
Glass
25 Pk.
F254
SiliaPlate
Bare Silica (Silicon Dioxyde, SiO2)
10 x 20 cm