Learn More
SiliCycle™ SiliaPlate™ TLC Plates, Glass-backed, Hard Layer Silica
Glass TLC plates exhibiting high chemical resistance, high heating stability and charring resistance for quick and inexpensive separations.
Supplier:  SiliCycle™ TLGR10014B124
 View more versions of this product
Catalog No. 01-184-495
Add to cart
Description
Description
SilicaPlate™ Glass-backed TLC Plates are made with irregular particle shape and exhibit high chemical resistance. A small percentage of inorganic, hardening agent has been added for a uniform and hard surface that will not crack, blister or swell up. These plates are designed for maximum robustness of the binder- they are very easy to handle and write on, and are completely wettable.
  • Rigid plates
  • High heating stability and charring resistance
  • Transparent
  • Hard layer silica
Specifications
Specifications
Glass
10 to 12 μm
Glass-backed TLC Plates
TLC Analysis
Bare Silica (Silicon Dioxyde, SiO2)
2.5 x 7.5 cm
Glass
100 Pk.
F254
SiliaPlate
250 μm