Learn More
SiliCycle™ SiliaPlate™ TLC Plates, Glass-backed, Silica, Optimized for KMnO4
Glass TLC plates exhibiting high chemical resistance, high heating stability and charring resistance for quick and inexpensive separations.
Supplier:  SiliCycle™ TLGR10014BK323N
 View more versions of this product
Catalog No. 01-184-445
Add to Cart
Description
Description
SilicaPlate™ Glass-backed TLC Plates are made with irregular particle shape and exhibit high chemical resistance. These plates are optimized for KMnO4.
  • Rigid plates
  • High heating stability and charring resistance
  • Transparent
  • Silica
Specifications
Specifications
Glass
10 to 12 μm
Glass-backed TLC Plates
TLC Analysis
Bare Silica (Silicon Dioxyde, SiO2)
20 x 20 cm
Glass
25 Pk.
No
SiliaPlate
250 μm