Learn More
SiliCycle™ SiliaPlate™ TLC Plates; Plastic-Backed, Silica
Plastic TLC plates for quick and inexpensive separations, complete with low fragility and ability to be cut by regular scissors.
Supplier:  SiliCycle™ TLPR10014BK0116
 View more versions of this product
Catalog No. 01-184-507
Add to Cart
Description
Description
SiliaPlate™ Plastic-backed TLC Plates are thin with irregular particle shape.
  • Low fragility
  • Possible to cut with scissors
  • High chemical resistance
  • Can be stored in notebook
Specifications
Specifications
Plastic
10 to 12 μm
Plastic-backed TLC Plates
TLC Analysis
Bare Silica (Silicon Dioxyde, SiO2)
3.3 x 6.6 cm
Silica 60
50 Pk.
F254
SiliaPlate
200 μm