Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Acidic Alumina
SPE Cartridges packed with Acidic Alumina, for extraction of aromatic compounds and aliphatic amines.
Supplier:  SiliCycle™ SPEAUT005306S
 View more versions of this product
Catalog No. 01-182-852
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Alumina
Extraction of aromatic compounds and aliphatic amines
75 to 150 μm
Acidic Alumina (Aluminum Oxide, Al2O3)
6 mL
Irregular
50 Pk.
12.7 x 77.8 mm
12.7 mm
1 g
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep™
SPE Cartridges
Polypropylene (PP) tube, Polyethylene (PE) frits
70 Å
150 to 320 m2/g
77.8 mm