Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - C1
SPE Cartridges packed with low hydrophobic and non-polar C1 sorbent (5% carbon), for extraction of highly hydrophobic pharmaceutical products.
$159.20 - $279.50
Specifications
Adsorbent Type Silica
Carbon Load 5%
For Use With (Application) Extraction of highly hydrophobic compounds
For Use With (Equipment) Vacuum Manifolds or Automated SPE Equipment
Particle Size 40 to 63 μm
View More Specs
Products 9
Catalog Number Mfr. No. Bed Weight Volume (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight Volume (Metric) Quantity Price Quantity & Availability  
01-184-676
View Documents
SiliCycle™
SPER33030B01B
50 mg 1 mL 100 Pk.
Pack of 100 for $239.00
Add to Cart
01-184-677
View Documents
SiliCycle™
SPER33030B01C
100 mg 1 mL 100 Pk.
Pack of 100 for $247.00
Add to Cart
01-184-678
View Documents
SiliCycle™
SPER33030B03G
200 mg 3 mL 50 Pk.
Pack of 50 for $159.20
Add to Cart
01-184-679
View Documents
SiliCycle™
SPER33030B03P
500 mg 3 mL 50 Pk.
Pack of 50 for $198.90
Add to Cart
01-184-680
View Documents
SiliCycle™
SPER33030B06P
500 mg 6 mL 50 Pk.
Pack of 50 for $198.90
Add to Cart
01-184-681
View Documents
SiliCycle™
SPER33030B06S
1 g 6 mL 50 Pk.
Pack of 50 for $216.50
Add to Cart
01-184-834
View Documents
SiliCycle™
SPER33030B06U
2 g 6 mL 50 Pk.
Pack of 50 for $253.00
Add to Cart
01-184-682
View Documents
SiliCycle™
SPER33030B12U
2 g 12 mL 20 Pk.
Pack of 20 for $228.50
Add to Cart
01-184-683
View Documents
SiliCycle™
SPER33030B20X
5 g 25 mL 20 Pk.
Pack of 20 for $279.50
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Extraction of highly hydrophobic compounds
40 to 63 μm
C1
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
5%
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep
SPE Cartridge
Irregular
SDS
Documents
Documents
Product Certifications
Keep dry