Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Carbonate
SPE Cartridges packed with the silica-bound equivalent of tetramethyl ammonium carbonate. Ideal for extraction of acids, for example scavenging of TFA
Supplier:  SiliCycle™ SPER66030B06U
 View more versions of this product
Catalog No. 01-184-586
Add to cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
2 g
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep™
SPE Cartridges
Yes
Irregular
50 Pk.
12.7 x 77.8 mm
12.7 mm
0.06
Scavenging of TFA and extraction of acids (boronic acids and acidic phenols)
40 to 63 μm
Carbonate (3-(Trimethylammonium)propyl)
6 mL
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
500 m2/g
77.8 mm