Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Carboxylic Acid (WCX) nec
SPE Cartridges packed with Carboxylic Acid sorbent (WCX), with no endcapping, for extraction of strong basic compounds.
$223.50 - $793.00
Specifications
Adsorbent Type Silica
For Use With (Application) Extraction of strong basic compounds
For Use With (Equipment) Vacuum Manifolds or Automated SPE Equipment
Particle Size 40 to 63 μm
Product Line SiliaPrep
View More Specs
Products 9
Catalog Number Mfr. No. Bed Weight Volume (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight Volume (Metric) Quantity Price Quantity & Availability  
01-183-389
View Documents
SiliCycle™
SPER70130B01B
50 mg 1 mL 100 Pk.
Pack of 100 for $263.50
Add to Cart
01-183-390
View Documents
SiliCycle™
SPER70130B01C
100 mg 1 mL 100 Pk.
Pack of 100 for $295.00
Add to Cart
01-183-391
View Documents
SiliCycle™
SPER70130B03G
200 mg 3 mL 50 Pk.
Pack of 50 for $223.50
Add to Cart
01-183-392
View Documents
SiliCycle™
SPER70130B03P
500 mg 3 mL 50 Pk.
Pack of 50 for $295.00
Add to Cart
01-183-393
View Documents
SiliCycle™
SPER70130B06P
500 mg 6 mL 50 Pk.
Pack of 50 for $310.50
Add to Cart
01-183-394
View Documents
SiliCycle™
SPER70130B06S
1 g 6 mL 50 Pk.
Pack of 50 for $414.50
Add to Cart
01-183-395
View Documents
SiliCycle™
SPER70130B06U
2 g 6 mL 50 Pk.
Pack of 50 for $793.00
Add to Cart
01-183-396
View Documents
SiliCycle™
SPER70130B12U
2 g 12 mL 20 Pk.
Pack of 20 for $344.00
Add to Cart
01-183-397
View Documents
SiliCycle™
SPER70130B20X
5 g 25 mL 20 Pk.
Pack of 20 for $714.00
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep
SPE Cartridge
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
Extraction of strong basic compounds
40 to 63 μm
Carboxylic Acid (WCX) nec
No
Irregular