Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Florisil LP
SPE Cartridges packed with Florisil 75 to 150 μm, for extraction of moderately polar compounds such as PCBs and polysaccharides.
Supplier:  SiliCycle™ SPEAUT0014LP12U
 View more versions of this product
Catalog No. 01-182-836
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Florisil
Extraction of chlorinated pesticides, PCBs and polysaccharides
75 to 150 μm
Florisil (Magnesium Silicate) LP
12 mL
Irregular
20 Pk.
15.8 x 90 mm
15.8 mm
2 g
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep™
SPE Cartridges
Polypropylene (PP) tube, Polyethylene (PE) frits
100 Å
250 m2/g
90 mm