Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Imidazole
SPE Cartridges packed with SiliaMetS Imidazole, to lower residual metal concentration of various metal complexes (Cd, Co, Cu, Fe, Li, Mg, Ni, Os & Zn)
Supplier:  SiliCycle™ SPER79230B12U
 View more versions of this product
Catalog No. 01-183-423
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Lower residual metal concentration in Cd, Co, Cu, Fe, Li, Mg, Ni, Os and Zn
40 to 63 μm
Imidazole
12 mL
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
15.8 x 90 mm
15.8 mm
2 g
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep
SPE Cartridge
Yes
Irregular
20 Pk.
90 mm
Product Suggestions
Product Suggestions