Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Imidazole
SPE Cartridges packed with SiliaMetS Imidazole, to lower residual metal concentration of various metal complexes (Cd, Co, Cu, Fe, Li, Mg, Ni, Os & Zn)
Supplier:  SiliCycle™ SPER79230B20X
 View more versions of this product
Catalog No. 01-183-424
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Lower residual metal concentration in Cd, Co, Cu, Fe, Li, Mg, Ni, Os and Zn
40 to 63 μm
Imidazole
25 mL
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
20.5 x 100 mm
20.5 mm
5 g
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep™
SPE Cartridges
Yes
Irregular
20 Pk.
100 mm