Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Pentafluorophenyl (PFP)
SPE Cartridges packed with moderately hydrophobic and highly polar PFP sorbent, for extraction of polar & aromatic compounds and separation of isomers
$295.00 - $1348.00
Specifications
Adsorbent Type Silica
Carbon Load 11%
For Use With (Application) Extraction of polar and aromatic compounds, separation of isomers
For Use With (Equipment) Vacuum Manifolds or Automated SPE Equipment
Particle Size 40 to 63 μm
View More Specs
Products 9
Catalog Number Mfr. No. Bed Weight Volume (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight Volume (Metric) Quantity Price Quantity & Availability  
01-183-344
View Documents
SiliCycle™
SPER67530B01B
50 mg 1 mL 100 Pk.
Pack of 100 for $350.00
Add to Cart
01-183-345
View Documents
SiliCycle™
SPER67530B01C
100 mg 1 mL 100 Pk.
Pack of 100 for $386.00
Add to Cart
01-183-346
View Documents
SiliCycle™
SPER67530B03G
200 mg 3 mL 50 Pk.
Pack of 50 for $295.00
Add to Cart
01-183-347
View Documents
SiliCycle™
SPER67530B03P
500 mg 3 mL 50 Pk.
Pack of 50 for $475.50
Add to Cart
01-183-348
View Documents
SiliCycle™
SPER67530B06P
500 mg 6 mL 50 Pk.
Pack of 50 for $516.50
Add to Cart
01-183-349
View Documents
SiliCycle™
SPER67530B06S
1 g 6 mL 50 Pk.
Pack of 50 for $768.50
Add to Cart
01-183-350
View Documents
SiliCycle™
SPER67530B06U
2 g 6 mL 50 Pk.
Pack of 50 for $1,348.00
Add to Cart
01-183-351
View Documents
SiliCycle™
SPER67530B12U
2 g 12 mL 20 Pk.
Pack of 20 for $785.50
Add to Cart
01-183-352
View Documents
SiliCycle™
SPER67530B20X
5 g 25 mL 20 Pk.
Pack of 20 for $1,315.00
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Extraction of polar and aromatic compounds, separation of isomers
40 to 63 μm
PFP (Pentafluorophenyl)
Yes
Irregular
500 m2/g
11%
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep
SPE Cartridge
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
SDS
Documents
Documents
Product Certifications
Keep dry