Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Piperazine
SPE Cartridges packed with SiliaBond Piperazine, for extraction of strong anions. Elution at lower pH (around 8 to 9) compared to SiliaPrep Amine (WAX).
Specifications
Adsorbent Type Silica
For Use With (Application) Extraction of strong anions (elution at lower pH - around 8 or 9 - compared to standard WAX)
For Use With (Equipment) Vacuum Manifolds or Automated SPE Equipment
Particle Size 40 to 63 μm
Product Line SiliaPrep
View More Specs
Products 9
Catalog Number Mfr. No. Bed Weight Volume (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight Volume (Metric) Quantity Price Quantity & Availability  
01-184-795
View Documents
SiliCycle™
SPER60030B01B
50 mg 1 mL 100 Pk. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
01-184-538
View Documents
SiliCycle™
SPER60030B01C
100 mg 1 mL 100 Pk. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
01-184-818
View Documents
SiliCycle™
SPER60030B03G
200 mg 3 mL 50 Pk. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
01-184-539
View Documents
SiliCycle™
SPER60030B03P
500 mg 3 mL 50 Pk. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
01-184-540
View Documents
SiliCycle™
SPER60030B06P
500 mg 6 mL 50 Pk. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
01-184-541
View Documents
SiliCycle™
SPER60030B06S
1 g 6 mL 50 Pk. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
01-184-850
View Documents
SiliCycle™
SPER60030B06U
2 g 6 mL 50 Pk. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
01-184-542
View Documents
SiliCycle™
SPER60030B12U
2 g 12 mL 20 Pk. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
01-184-543
View Documents
SiliCycle™
SPER60030B20X
5 g 25 mL 20 Pk. Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep
SPE Cartridge
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
Extraction of strong anions (elution at lower pH - around 8 or 9 - compared to standard WAX)
40 to 63 μm
Piperazine
Yes
Irregular