Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - TAAcONa
SPE Cartridges packed with SiliaMetS TAAcONa, to lower residual metal concentration of major metal complexes (Ca, Cd, Cu, Fe, Mg, Ni, Os, Rh, Sc & Sn)
Supplier:  SiliCycle™ SPER69230B01C
 View more versions of this product
Catalog No. 01-183-363
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Lower residual metal concentration in Ca, Cd, Cu, Fe, Mg, Ni, Os, Rh, Sc and Sn
40 to 63 μm
TAAcONa
1 mL
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
5.7 x 65.7 mm
5.7 mm
100 mg
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep
SPE Cartridge
No
Irregular
100 Pk.
65.7 mm
Recommendations
Recommendations