Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Thiourea
SPE Cartridges packed with SiliaMetS Thiourea, to lower residual metal concentration of various metal complexes (Pd & Ru).
$180.60 - $335.00
Specifications
Adsorbent Type Silica
For Use With (Application) Lower residual metal concentration in Pd and Ru
For Use With (Equipment) Vacuum Manifolds or Automated SPE Equipment
Particle Size 40 to 63 μm
Product Line SiliaPrep
View More Specs
Products 9
Catalog Number Mfr. No. Bed Weight Volume (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight Volume (Metric) Quantity Price Quantity & Availability  
01-183-371
View Documents
SiliCycle™
SPER69530B01B
50 mg 1 mL 100 Pk.
Pack of 100 for $239.00
Add to Cart
01-183-372
View Documents
SiliCycle™
SPER69530B01C
100 mg 1 mL 100 Pk.
Pack of 100 for $239.00
Add to Cart
01-183-373
View Documents
SiliCycle™
SPER69530B03G
200 mg 3 mL 50 Pk.
Pack of 50 for $180.60
Add to Cart
01-183-374
View Documents
SiliCycle™
SPER69530B03P
500 mg 3 mL 50 Pk.
Pack of 50 for $198.90
Add to Cart
01-183-375
View Documents
SiliCycle™
SPER69530B06P
500 mg 6 mL 50 Pk.
Pack of 50 for $208.50
Add to Cart
01-183-376
View Documents
SiliCycle™
SPER69530B06S
1 g 6 mL 50 Pk.
Pack of 50 for $256.50
Add to Cart
01-183-377
View Documents
SiliCycle™
SPER69530B06U
2 g 6 mL 50 Pk.
Pack of 50 for $335.00
Add to Cart
01-183-378
View Documents
SiliCycle™
SPER69530B12U
2 g 12 mL 20 Pk.
Pack of 20 for $208.50
Add to Cart
01-183-379
View Documents
SiliCycle™
SPER69530B20X
5 g 25 mL 20 Pk.
Pack of 20 for $270.50
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep
SPE Cartridge
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
Lower residual metal concentration in Pd and Ru
40 to 63 μm
Thiourea
Yes
Irregular