Learn More
SiliCycle™ SiliaPrep™ SPE Cartridges - Tridecafluoro
SPE Cartridges packed with Tridecafluoro sorbent, for fast and easy separation of fluorine containing molecules from non-fluorous ones.
Supplier:  SiliCycle™ SPER63530B06P
 View more versions of this product
Catalog No. 01-184-574
Add to Cart
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Separation of fluorine containing molecules from non-fluorous ones
40 to 63 μm
Tridecafluoro (TDF)
6 mL
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
12.7 x 77.8 mm
12.7 mm
500 mg
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep
SPE Cartridge
Yes
Irregular
50 Pk.
77.8 mm
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Keep dry