Learn More
SiliCycle™ SiliaPrepX™ SPE Cartridges - HLB
SPE Cartridges packed with wettable copolymer presenting a Hydrophilic-Lipophilic Balance (HLB), for extraction of neutral, acidic or basic compounds.
$183.00 - $2699.00
Specifications
Adsorbent Type | Polymer |
---|---|
Carbon Load | 0.88 |
For Use With (Application) | Extraction of drugs and metabolites from biological fluids, API from tablets and cream |
For Use With (Equipment) | Vacuum Manifolds or Automated SPE Equipment |
Particle Size | 40 μm |
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
01-182-924
|
SiliCycle™
SPEP000201AA |
30 mg | 1 mL | 100 Pk. |
Pack of 100 for $246.00
|
|
01-182-925
|
SiliCycle™
SPEP000201AAL |
30 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,282.00
|
|
01-182-926
|
SiliCycle™
SPEP000201AAM |
30 mg | 1 mL | 1000 Pk. |
Pack of 1000 for $2,699.00
|
|
01-182-930
|
SiliCycle™
SPEP000203AA |
30 mg | 3 mL | 50 Pk. |
Pack of 50 for $183.00
|
|
01-182-931
|
SiliCycle™
SPEP000203AAJ |
30 mg | 3 mL | 200 Pk. |
Pack of 200 for $655.00
|
|
01-182-927
|
SiliCycle™
SPEP000201BB |
60 mg | 1 mL | 100 Pk. |
Pack of 100 for $278.00
|
|
01-182-928
|
SiliCycle™
SPEP000201BBL |
60 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,320.00
|
|
01-182-932
|
SiliCycle™
SPEP000203BB |
60 mg | 3 mL | 50 Pk. |
Pack of 50 for $217.00
|
|
01-182-933
|
SiliCycle™
SPEP000203BBJ |
60 mg | 3 mL | 200 Pk. |
Pack of 200 for $823.00
|
|
01-182-937
|
SiliCycle™
SPEP000206BB |
60 mg | 6 mL | 30 Pk. |
Pack of 30 for $204.00
|
|
01-182-938
|
SiliCycle™
SPEP000206BBJ |
60 mg | 6 mL | 200 Pk. |
Pack of 200 for $1,012.00
|
|
01-182-934
|
SiliCycle™
SPEP000203C |
100 mg | 3 mL | 50 Pk. |
Pack of 50 for $231.00
|
|
01-182-935
|
SiliCycle™
SPEP000203CJ |
100 mg | 3 mL | 200 Pk. |
Pack of 200 for $876.00
|
|
01-182-936
|
SiliCycle™
SPEP000203CL |
100 mg | 3 mL | 500 Pk. |
Pack of 500 for $1,957.00
|
|
01-182-939
|
SiliCycle™
SPEP000206C |
100 mg | 6 mL | 30 Pk. |
Pack of 30 for $217.00
|
|
01-182-940
|
SiliCycle™
SPEP000206G |
200 mg | 6 mL | 30 Pk. |
Pack of 30 for $246.00
|
|
01-182-941
|
SiliCycle™
SPEP000206GJ |
200 mg | 6 mL | 200 Pk. |
Pack of 200 for $1,222.00
|
|
01-182-942
|
SiliCycle™
SPEP000212P |
500 mg | 12 mL | 20 Pk. |
Pack of 20 for $302.00
|
|
01-182-923
|
SiliCycle™
SPEP0002011A |
10 mg | 1 mL | 100 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
01-182-929
|
SiliCycle™
SPEP0002031A |
10 mg | 3 mL | 50 Pk. | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- For use with vacuum manifolds and automated SPE equipments
- Tightly packed and homogeneous silica bed for reliability and increased productivity
- Tight particle size distribution and very good packing with no fines, allowing excellent separation
- High recoveries and yields
- Reproducible flow rates lot-to-lot
- Flanged polypropylene tubes
- 20 μm polyethylene frits
Specifications
Polymer | |
Extraction of drugs and metabolites from biological fluids, API from tablets and cream | |
40 μm | |
HLB (Hydrophilic-Lipophilic Balance) | |
Polypropylene (PP) tube, Polyethylene (PE) frits | |
110 Å |
0.88 | |
Vacuum Manifolds or Automated SPE Equipment | |
SiliaPrepX™ | |
SPE Cartridges | |
Spherical | |
850 m2/g |