Learn More
SiliCycle™ SiliaPrepX™ SPE Cartridges - SAX
SPE Cartridges packed with a polymeric strong anion exchange (SAX) phase, for extraction of weak & medium acids.
$192.20 - $2450.00
Specifications
Adsorbent Type | Polymer |
---|---|
Carbon Load | 0.85 |
For Use With (Application) | Extraction of weak and medium acids, highly stable in organic solvents |
For Use With (Equipment) | Vacuum Manifolds or Automated SPE Equipment |
Particle Size | 85 μm |
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Bed Weight | Volume (Metric) | Quantity | Price | Quantity & Availability | |||
01-182-974
|
SiliCycle™
SPEP001001AA |
30 mg | 1 mL | 100 Pk. |
Pack of 100 for $272.00
|
|
|||
01-182-975
|
SiliCycle™
SPEP001001AAL |
30 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,289.00
|
|
|||
01-182-976
|
SiliCycle™
SPEP001001AAM |
30 mg | 1 mL | 1000 Pk. |
Pack of 1000 for $2,450.00
|
|
|||
01-182-981
|
SiliCycle™
SPEP001003AA |
30 mg | 3 mL | 50 Pk. |
Pack of 50 for $192.20
|
|
|||
01-182-982
|
SiliCycle™
SPEP001003AAJ |
30 mg | 3 mL | 200 Pk. |
Pack of 200 for $688.00
|
|
|||
01-182-977
|
SiliCycle™
SPEP001001BB |
60 mg | 1 mL | 100 Pk. |
Pack of 100 for $292.00
|
|
|||
01-182-978
|
SiliCycle™
SPEP001001BBL |
60 mg | 1 mL | 500 Pk. |
Pack of 500 for $1,386.00
|
|
|||
01-182-983
|
SiliCycle™
SPEP001003BB |
60 mg | 3 mL | 50 Pk. |
Pack of 50 for $228.00
|
|
|||
01-182-984
|
SiliCycle™
SPEP001003BBJ |
60 mg | 3 mL | 200 Pk. |
Pack of 200 for $864.50
|
|
|||
01-182-990
|
SiliCycle™
SPEP001006BB |
60 mg | 6 mL | 30 Pk. |
Pack of 30 for $206.00
|
|
|||
01-182-991
|
SiliCycle™
SPEP001006BBJ |
60 mg | 6 mL | 200 Pk. |
Pack of 200 for $1,026.00
|
|
|||
01-182-979
|
SiliCycle™
SPEP001001C |
100 mg | 1 mL | 100 Pk. |
Pack of 100 for $338.50
|
|
|||
01-182-985
|
SiliCycle™
SPEP001003C |
100 mg | 3 mL | 50 Pk. |
Pack of 50 for $243.00
|
|
|||
01-182-986
|
SiliCycle™
SPEP001003CJ |
100 mg | 3 mL | 200 Pk. |
Pack of 200 for $920.00
|
|
|||
01-182-987
|
SiliCycle™
SPEP001003CL |
100 mg | 3 mL | 500 Pk. |
Pack of 500 for $2,180.00
|
|
|||
01-182-992
|
SiliCycle™
SPEP001006C |
100 mg | 6 mL | 30 Pk. |
Pack of 30 for $258.50
|
|
|||
01-182-988
|
SiliCycle™
SPEP001003G |
200 mg | 3 mL | 50 Pk. |
Pack of 50 for $287.00
|
|
|||
01-182-993
|
SiliCycle™
SPEP001006G |
200 mg | 6 mL | 30 Pk. |
Pack of 30 for $272.00
|
|
|||
01-182-989
|
SiliCycle™
SPEP001003P |
500 mg | 3 mL | 50 Pk. |
Pack of 50 for $513.50
|
|
|||
01-182-994
|
SiliCycle™
SPEP001006P |
500 mg | 6 mL | 30 Pk. |
Pack of 30 for $367.50
|
|
|||
01-182-996
|
SiliCycle™
SPEP001012P |
500 mg | 12 mL | 20 Pk. |
Pack of 20 for $317.50
|
|
|||
01-182-995
|
SiliCycle™
SPEP001006S |
1 g | 6 mL | 30 Pk. |
Pack of 30 for $434.00
|
|
|||
01-182-998
|
SiliCycle™
SPEP001020S |
1 g | 25 mL | 20 Pk. |
Pack of 20 for $610.50
|
|
|||
01-182-997
|
SiliCycle™
SPEP001012U |
2 g | 12 mL | 20 Pk. |
Pack of 20 for $756.00
|
|
|||
01-182-999
|
SiliCycle™
SPEP001020U |
2 g | 25 mL | 20 Pk. |
Pack of 20 for $727.00
|
|
|||
01-182-973
|
SiliCycle™
SPEP0010011A |
10 mg | 1 mL | 100 Pk. | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||
01-182-980
|
SiliCycle™
SPEP0010031A |
10 mg | 3 mL | 50 Pk. | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||
Description
- For use with vacuum manifolds and automated SPE equipments
- Tightly packed and homogeneous silica bed for reliability and increased productivity
- Tight particle size distribution and very good packing with no fines, allowing excellent separation
- High recoveries and yields
- Reproducible flow rates lot-to-lot
- Flanged polypropylene tubes
- 20 μm polyethylene frits
Specifications
Polymer | |
Extraction of weak and medium acids, highly stable in organic solvents | |
85 μm | |
Polystyrene-divinylbenzene SAX | |
Polypropylene (PP) tube, Polyethylene (PE) frits | |
60 Å |
0.85 | |
Vacuum Manifolds or Automated SPE Equipment | |
SiliaPrepX™ | |
SPE Cartridges | |
Spherical | |
900 m2/g |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.