Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SiliCycle™ SiliaSphere™ C18, 10 μm, 100 Å
SiliaSphere™ C18, 10 μm, 100 Å is a great start for method development as it presents the maximum retention of non-polar compounds.
Supplier: SiliCycle™ S03207EA100G
Description
- Usually used for peptides, pesticides, PCBs, PAHs, drugs, etc.
Specifications
10 μm | |
C18 | |
Bulk | |
C18 (Octadecyl, ODS) | |
Bulk Functionalized Spherical Silica Gel | |
High Pressure Chromatography | |
Yes | |
Spherical |
100 Å | |
Liquid Chromatography | |
100 g | |
SiliaSphere™ | |
Silica Gel | |
Separation of low to medium polarity compounds | |
Reversed-phase |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title