Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SiliCycle™ SiliaSphere™ C18, 5 μm, 100 Å
SiliaSphere™ C18, 5 μm, 100 Å is a great start for method development as it presents the maximum retention of non-polar compounds.
$168.00 - $9340.00
Specifications
Particle Size | 5 μm |
---|---|
Pore Size | 100 Å |
Stationary Phase | C18 |
Technique | Liquid Chromatography |
Packaging | Bulk |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-184-319
|
SiliCycle™
S03205EA5G |
5 g |
Each for $168.00
|
|
01-184-315
|
SiliCycle™
S03205EA10G |
10 g |
Each for $309.00
|
|
01-184-349
|
SiliCycle™
S03205EA25G |
25 g |
Each for $707.00
|
|
01-184-318
|
SiliCycle™
S03205EA50G |
50 g |
Each for $1,284.00
|
|
01-184-314
|
SiliCycle™
S03205EA100G |
100 g |
Each for $2,510.00
|
|
01-184-316
|
SiliCycle™
S03205EA250G |
250 g |
Each for $5,770.00
|
|
01-184-317
|
SiliCycle™
S03205EA500G |
500 g |
Each for $9,340.00
|
|
Description
- Usually used for peptides, pesticides, PCBs, PAHs, drugs, etc.
Specifications
5 μm | |
C18 | |
Bulk | |
SiliaSphere™ | |
Silica Gel | |
Separation of low to medium polarity compounds | |
Reversed-phase |
100 Å | |
Liquid Chromatography | |
C18 (Octadecyl, ODS) | |
Bulk Functionalized Spherical Silica Gel | |
High Pressure Chromatography | |
Yes | |
Spherical |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title