Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaSphere™ C18, 5 μm, 300 Å

SiliaSphere™ C18, 5 μm, 300 Å is a great start for method development as it presents the maximum retention of non-polar compounds.

Supplier:  SiliCycle™ S03205M100G

Request Bulk Order

 View more versions of this product

Catalog No. 01-184-320


Explore available promotions
Add to Cart

Description

Description

  • Usually used for peptides, pesticides, PCBs, PAHs, drugs, etc.
Specifications

Specifications

5 μm
C18
Bulk
C18 (Octadecyl, ODS) WPD
Bulk Functionalized Spherical Silica Gel
High Pressure Chromatography
Yes
Spherical
300 Å
Liquid Chromatography
100 g
SiliaSphere™
Silica Gel
Separation of low to medium polarity compounds
Reversed-phase
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.