Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaSphere™ PC Spherical Silica Gel, 20 to 45 μm, 300 Å

SiliaSphere™ PC Spherical Silica Gel. Increased efficiency of eluent’s flow characteristics, improved resolution between sample compounds and ease of packing.

Supplier:  SiliCycle™ S10020M500G

 View more versions of this product

Catalog No. 01-182-532


Add to Cart

Description

Description

SiliaSphere™ PC - Bulk Spherical Silica Gels for Preparative Chromatography

In preparative and process chromatography, the cost is very important and the use of spherical particles with narrow particle size distribution is very expensive. It is possible in this case to use irregular or angular silica but the separation will not be as optimal.

SiliCycle™ has added a very efficient material for preparative chromatography, the SiliaSphere™ PC spherical silica gels. These products have a very high chemical and mechanical stability. They also have the advantage to operate with very low backpressures. Furthermore, they are being offered at a very affordable price.

Advantages of using the SiliaSphere™ PC materials over standard silica gels:

  • Increased efficiency of the eluent's flow characteristics
  • Better resolution
  • Ease of packing
  • Very high specific surface area
  • High mechanical stability under higher pressures

Various pore sizes are available in a wide range of particle size (20 to 200 μm).

Specifications

Specifications

20 to 45 μm
Silica
Bulk
Bare Silica (Silicon Dioxyde, SiO2)
SiliaSphere™ PC
Silica Gel
7631-86-9
300 Å
Column Chromatography
500 g
Keep dry
Bulk Spherical Silica Gel
Chromatography, purification
Spherical
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Keep dry