Learn More
SiliCycle™ SiliaSphere™ PC Spherical Silica Gel, 75 to 200 μm, 1,000 Å
SiliaSphere™ PC Spherical Silica Gel. Increased efficiency of eluent’s flow characteristics, improved resolution between sample compounds and ease of packing.
Supplier: SiliCycle™ S10040T500G
Description
SiliaSphere™ PC - Bulk Spherical Silica Gels for Preparative Chromatography
In preparative and process chromatography, the cost is very important and the use of spherical particles with narrow particle size distribution is very expensive. It is possible in this case to use irregular or angular silica but the separation will not be as optimal.
SiliCycle™ has added a very efficient material for preparative chromatography, the SiliaSphere™ PC spherical silica gels. These products have a very high chemical and mechanical stability. They also have the advantage to operate with very low backpressures. Furthermore, they are being offered at a very affordable price.
Advantages of using the SiliaSphere™ PC materials over standard silica gels:
- Increased efficiency of the eluent's flow characteristics
- Better resolution
- Ease of packing
- Very high specific surface area
- High mechanical stability under higher pressures
Various pore sizes are available in a wide range of particle size (20 to 200 μm).
Specifications
75 to 200 μm | |
Silica | |
Bulk | |
Bare Silica (Silicon Dioxyde, SiO2) | |
SiliaSphere™ PC | |
Silica Gel | |
7631-86-9 | |
Spherical |
1000 Å | |
Column Chromatography | |
500 g | |
Keep dry | |
Bulk Spherical Silica Gel | |
Chromatography, purification | |
No |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Keep dry