Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaSphere™ PC Spherical Silica Gel, C18 Monomeric, 50 μm, 60 Å

SiliaSphere™ PC Spherical Silica Gel, C18 Monomeric, 50 μm, 60 Å is a C18 phase, a great start for reversed-phase method development, as it presents the maximum retention of non-polar compounds.

Supplier:  SiliCycle™ S03230BA1KG

 View more versions of this product

Catalog No. 01-182-375


Add to Cart

Description

Description

In preparative and process chromatography the cost is very important and the use of spherical particles with narrow particle size distribution is very expensive. It is possible in this case to use irregular or angular silica but the separation will not be as optimal. SiliCycle™ has added a very efficient material for preparative chromatography, the SiliaSphere™ PC spherical silica gels. These products have a very high chemical and mechanical stability. They also have the advantage to operate with very low backpressures. Furthermore, they are being offered at a very affordable price.

Advantages of using the SiliaSphere™ PC materials over standard silica gels:

  • Increased efficiency of the eluent's flow characteristics
  • Better resolution
  • Ease of packing
  • Very high specific surface area
  • High mechanical stability under higher pressures
  • Usually used for peptides, pesticides, PCBs, PAHs, drugs, etc.
  • Various pore sizes are available in a wide range of particle size (20 to 200 μm)
Specifications

Specifications

50 μm
C18
Bulk
C18 (Octadecyl, ODS)
Bulk Functionalized Spherical Silica Gel
Glass Gravity Columns
Yes
Spherical
60 Å
Column Chromatography
1 kg
SiliaSphere™ PC
Silica Gel
Separation of low to medium polarity compounds
Reversed-phase
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit