Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SiliCycle™ SiliaSphere™ Spherical Silica Gel, 10 μm, 100 Å

Highly polar sorbent for the purification of non-polar and non-ionic compounds.

Supplier:  SiliCycle™ S10007EA5G

 View more versions of this product

Catalog No. 01-184-348


Add to cart

Description

Description

SiliaSphere- Bulk Analytical Spherical Silica Gels

SiliaSphere, the right choice for:

  • Chromatographic performance
  • Loading capacity
  • Reproducibility
  • Chemical & physical stability

With pore diameters ranging from 60 to 1000 Å and particle sizes from 1.8 to 25 μm, we SiliCycle™ products to meet all your separation requirements. This is one of the most reliable portfolios of spherical silica gels for high pressure chromatography.

SiliaSphere are ideal for both analytical and preparative chromatography, from laboratory to pilot-plant processes and production scales. Furthermore, the excellent properties of SiliaSphere make them the packing of choice for high performance liquid chromatography (HPLC), Supercritical Fluid Chromatography (SFC) and simulated moving bed (SMB) applications.

Features and Benefits of SiliaSphere™ Spherical Silica Gels:

  • High purity silica gels (Consistency, reliability & reproducibility)
  • Perfect spherical shape, free of any cavities or cracks (Ease of column packing and high resolution)
  • Exceptional narrow particle size distribution (Optimal separation and resolution)
  • Strong mechanical stability (Low back-pressure without surface abrasion)
  • Same well controlled processes for all SiliaSphere (Easy scalability)

SiliaSphere as a Silica Matrix

SiliCycle has a strong know-how and expertise in silica gel manufacturing. To support the increasing demand on spherical silicas, SiliCycle developed an optimized and highly controlled large-scale production process for all of their SiliaSphere products, without decreasing the quality of the silica.

Particle shape, pore & particle size distributions, silica gel purity and surface properties, all have their influence on chromatographic performance. Therefore, in order to develop the most efficient process, all these parameters need to be evaluated and optimized to ensure batch-to-batch reproducibility.

SiliaSphere is manufactured from an organic form of silicon (alkoxydes). This ensures very low metal content as the starting material is purified by distillation. Deionized water is used to hydrolyze the silicon alkoxydes. Careful monitoring and control of the parameters that induce precipitation provide spherical silica gels with the desired characteristics.

SiliaSphere products are characterized by a very low metal content and exceptional stability. Furthermore, SiliCycle™'s manufacturing process ensures quality and reproducibility in pore size, surface area, particle size and morphology for all SiliaSphere products.

Specifications

Specifications

10 μm
Silica
Bulk
Bare Silica (Silicon Dioxyde, SiO2)
SiliaSphere™
Silica Gel
7631-86-9
Spherical
100 Å
Liquid Chromatography
5 g
Keep dry
Bulk Spherical Silica Gel
Chromatography, purification
No
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Keep dry