Learn More
Silica gel 60, 0.062-0.105mm (150-230 mesh), Thermo Scientific Chemicals
Description
Silica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | -150+230 Mesh,S.A. 500-600m |
Material | Silica Gel |
Quantity | 250 g |
Mesh Size | 150-230 Mesh |
Odor | Odorless |
Particle Size | 0.062 to 0.105mm |
Solubility Information | Soluble in hydrofluoric acid. Insoluble in water. |
Surface Area | 500 to 600m2/g |
Chemical Name or Material | Silica gel 60 |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H373 May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | GHS P Statement P260-P314-P501a Do not breathe dust/fume/gas/mist/vapors/spray. Get medical advice/attention if you feel unwell. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 231-545-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.