Silica gel 60, Alfa Aesar™

AA4272918

Manufacturer: Alfa Aesar 42729A7

Quantity 5kg

 View more versions of this product

Catalog No. AA42729A7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7631-86-9
    Physical Form Powder
    MDL Number MFCD00011232
    Chemical Name or Material Silica gel 60

    Specifications

    Physical Form Powder
    Quantity 5kg
    Color White
    EINECS Number 231-545-4
    Appearance White powder
    MDL Number MFCD00011232
    CAS 7631-86-9
    Odor Odorless
    Health Hazard 1 H373: May cause damage to organs through prolonged or repeated exposure.
    Chemical Name or Material Silica gel 60

    H373: May cause damage to organs through prolonged or repeated exposure.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit