Learn More
MilliporeSigma™ Silica gel 60 PF254 for preparative thin layer chromatography, MilliporeSigma™
Silica gel 60 PF254 for preparative thin layer chromatography. CAS No. 7631-86-9, EC Number 231-545-4.
$191.80 - $395.50
Specifications
Grade 60 PF254
Material Silica Gel
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M1077471000
View Documents
MilliporeSigma™
1.07747.1000
1 kg
Each for $191.80
Add to cart
M1077472500
View Documents
MilliporeSigma™
1.07747.2500
2.5 kg
Each for $395.50
Add to cart
Description
Description
Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40µm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).
Specifications
Specifications
60 PF254
Silica Gel