Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica gel, C8-RP, 12%C, ca. 1.2mmol/g, part. size 40-63μ, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
MFCD00148339
Octylsilicate
MFCD00148339
Octylsilicate
For column chromatography
Loading 1.21mmol/g min.
Elemental Analysis 11.6% min. (Carbon)
Color White
Quantity 250g
Packaging Plastic bottle
Particle Size 40 to 63 Micron
Physical Form Powder
Volatiles 4% max. (110°C)
Chemical Name or Material Silica gel, C8-RP, 12%C, ca. 1.2 mmol/g, part. size 40-63 micron