Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica gel, for column chrom., ultrapure, 60-200μ, 60A, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic Bottle
Plastic drum
7631-86-9
O2Si
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Silicon dioxide
24261
CHEBI:30563
dioxosilane
O=[Si]=O
7631-86-9
O2Si
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Silicon dioxide
24261
CHEBI:30563
dioxosilane
O=[Si]=O
For column chromatography
Color White to Brown
pH 6.5 to 7.5 (10% w/w suspension)
Quantity 250g
Pore Volume 0.70 to 0.85mL/g (specific)
Packaging Plastic Bottle
Particle Size 5.0% Max. (under size) 5.0% Max. (over size)
Solubility Information Solubility in water: insoluble.
Specific Surface Area 480 to 550 m2/g (BET)
Formula Weight 60.08
Physical Form Powder and/or Chunks
Volatiles 3.0 to 6.0 % (160°C)
Chemical Name or Material Silica gel, For column chromatography
Show More Show Less