Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel, Davisil(R), Grade 635, 60-100 Mesh, 60A, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
Steel Can
7631-86-9
O2Si
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
O=[Si]=O
7631-86-9
O2Si
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
O=[Si]=O

SIL63, 7631-86-9

Spectrum™ Silica Gel, Davisil(R), Grade 635, 60-100 Mesh, 60A , a form of silicon dioxide, is made from sodium silicate. As a granular and porous solid, it is a naturally occurring mineral and often used as a desiccant to control humidity and avoid spoilage. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

CAS Max % 1
Quantity 1 kg
Packaging Steel Can
Particle Size 60-100 Mesh
Grade Reagent