Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica gel, for drying purposes, non-toxic grade, 3-6 mm, ACROS Organics™

Click to view available options
Quantity:
1kg
5kg
25kg
Packaging:
Fibre drum
Plastic Bottle
Plastic drum
7631-86-9
O2Si
60.083
MFCD00011232
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Silicon dioxide
24261
CHEBI:30563
dioxosilane
O=[Si]=O
7631-86-9
O2Si
60.083
MFCD00011232
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Silicon dioxide
24261
CHEBI:30563
dioxosilane
O=[Si]=O
Drying agent, For column chromatography
Bulk Density 620 to 800g/L
Additional Information 2.0% max. (140°C, 4 hrs)
Adsorption Adsorption capacity (relative humidity 40%): ca 20%
Color White to Yellow
Quantity 5kg
Packaging Plastic drum
Moisture Residual moisture (140°C): < 1% (typically)
Merck Index 15, 8632
Particle Size Particle size analysis: typically sieve residue>5mm:ca 10%, passing through 2mm: ca 1%
Solubility Information Solubility in water: insoluble.
Formula Weight 60.08
Physical Form Solid
Grade Non-toxic
Water Absorption 20.0% min. (40% min. RH)
Chemical Name or Material Silica gel, 3-6mm
Show More Show Less

EINECSNumber : 231-545-4

RTECSNumber : EU8655000

TSCA : TSCA