Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica gel, functionalized, diethylenetriamine tetraacetate, 0.5mmol/g, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7631-86-9
O2Si
60.083
MFCD06660072
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
dioxosilane
O=[Si]=O
7631-86-9
O2Si
60.083
MFCD06660072
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
dioxosilane
O=[Si]=O
For solid support synthesis
Loading 0.5mmol/g, exact loading on the certificate of analysis
Packaging Glass bottle
Quantity 1g
Color Yellow
Merck Index 15, 8632
Formula Weight 60.08
Physical Form Powder
Chemical Name or Material Silica gel, functionalized, diethylenetriamine tetraacetate, Functionalized, Diethylenetriamine tetraacetate, 0.5 mmol/g

GHS H Statement:
Toxic if inhaled.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.