Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel, Grade 633, 200-425 Mesh, 60A, Spectrum™

Click to view available options
Quantity:
100 g
5 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
Steel Can
7631-86-9
O2Si
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
O=[Si]=O
7631-86-9
O2Si
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
O=[Si]=O

SIL61, 7631-86-9

Spectrum™ Silica Gel, Grade 633, 200-425 Mesh, 60A is a form of silicon dioxide made from sodium silicate. It is granular and porous solid and is a naturally occurring mineral. It is often used as a desiccant to control humidity and avoid spoilage. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

CAS Max % 1
Quantity 5 kg
Pore Volume 0.70-0.85 CC/G
Packaging Poly Pail
Surface Area 480 m2/g
Grade Reagent