Alfa Aesar™ Silica gel, HPLC grade, spherical, 5 micron APS, 260 angstroms, 99.99+%

Manufacturer: Alfa Aesar™ 4581214

Quantity 25g

 View more versions of this product

Catalog No. AA4581214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63231-67-4
    MDL Number MFCD00011232
    Chemical Name or Material Silica gel, P.V. 0.90cc/g
    Grade HPLC Grade

    Specifications

    Health Hazard 3 GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 63231-67-4
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H373-H319-H335
    May cause damage to organs through prolonged or repeated exposure.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    Physical Form Spherical
    MDL Number MFCD00011232
    Surface Area 150m2/g
    RTECS Number VV7340000
    Quantity 25g
    Particle Size 3 Micron APS, 260Å
    Chemical Name or Material Silica gel, P.V. 0.90cc/g
    Assay ≥99.99%
    Grade HPLC Grade

    GHS H Statement
    H373-H319-H335
    May cause damage to organs through prolonged or repeated exposure.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit