Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silica gel, preparative chromatography grade, spherical, 20 micron APS, 100 angstroms, 99.99+%

Manufacturer:  Alfa Aesar™ 4583122

Quantity 100g

 View more versions of this product

Catalog No. AA4583122


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

63231-67-4
MFCD00011232
Silica gel, P.V. 1.00cc/g
Preparative Chromatography Grade

Specifications

100g
20 Micron APS, 100Å
MFCD00011232
Silica gel, P.V. 1.00cc/g
Preparative Chromatography Grade
63231-67-4
Spherical
335m2/g
≥99.99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H373-H319-H335
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VV7340000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit