Learn More
Alfa Aesar™ Silica gel, spherical, 300 angstroms
Manufacturer:  Alfa Aesar™ 4409909
Quantity 10g
 View more versions of this product
Catalog No. AA4409909
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
63231-67-4
MFCD00011232
Silica gel, P.V. 0.9cc/g
Specifications
10g
5 Micron APS, 300Å
MFCD00011232
Silica gel, P.V. 0.9cc/g
63231-67-4
Spherical
100m2/g
Safety and Handling
SDS & Safety Documents
GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA