Alfa Aesar™ Silica gel, spherical, 60 angstroms

column chromatography

Manufacturer: Alfa Aesar™ 4409518

Quantity 50g

 View more versions of this product

Catalog No. AA4409518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63231-67-4
    MDL Number MFCD00011232
    Chemical Name or Material Silica gel, P.V. 0.7cc/g

    Specifications

    Health Hazard 1 Warning
    TSCA TSCA
    Physical Form Spherical
    MDL Number MFCD00011232
    CAS 63231-67-4
    Health Hazard 2 GHS H Statement
    H373
    May cause damage to organs through prolonged or repeated exposure.
    Quantity 50g
    Particle Size 5 Micron APS, 60Å
    Surface Area 450m2/g
    Health Hazard 3 GHS P Statement
    P260-P314-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Silica gel, P.V. 0.7cc/g

    GHS H Statement
    H373
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P314-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit