Learn More
Alfa Aesar™ Silica gel, TLC high purity grade, with gypsum binder
TLC plate preparation
Manufacturer:  Alfa Aesar™ 42763A1
Quantity 1kg
 View more versions of this product
Catalog No. AA42763A1
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
63231-67-4
MFCD00011232
Silica gel, With gypsum binder
TLC High Purity Grade
Specifications
1kg
63231-67-4
MFCD00011232
Silica gel, With gypsum binder
6 to 7
500 to 600m2/g, 60Å
500 to 600m2/g
TLC High Purity Grade
Safety and Handling
SDS & Safety Documents
GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA