Alfa Aesar™ Silica gel, TLC high purity grade, with gypsum binder and fluorescent indicator

TLC plate preparation

Manufacturer: Alfa Aesar™ 4276418

Quantity 50g

 View more versions of this product

Catalog No. AA4276418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63231-67-4
    MDL Number MFCD00011232
    Chemical Name or Material Silica gel, With gypsum binder and fluorescent indicator
    Grade TLC High Purity Grade

    Specifications

    CAS 63231-67-4
    MDL Number MFCD00011232
    Surface Area 500 to 600m2/g
    Health Hazard 3 GHS P Statement
    P260-P314-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 50g
    TSCA TSCA
    Particle Size 500 to 600m2/g, 60Å
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H373
    May cause damage to organs through prolonged or repeated exposure.
    pH 6.5 to 7.5
    Chemical Name or Material Silica gel, With gypsum binder and fluorescent indicator
    Grade TLC High Purity Grade

    GHS H Statement
    H373
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P314-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit